missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2,2-Tetrabromoethane-d2, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1,1,2,2-Tetrabromoethane-D2 |
| CAS | 33685-55-1 |
| Molecular Formula | C2 D2 Br4 |
| Quantity | 0.1 g |
| Synonym | Ethane-1,2-d2, 1,1,2,2-tetrabromo- (8 CI, 9 CI), Ethane-1,2-d2, tetrabromo- (6 CI), 1,1,2,2-Tetrabromoethane-d2, 1,1,2,2-Tetrabromoethane-D2, Tetrabromoethane-1,2-d2 |
| InChI Formula | InChI=1 S/C2H2Br4/c3-1(4)2(5)6/h1-2 H/i1D,2 D |
| SMILES | [2 H]C(Br)(Br)C([2 H])(Br)Br |
| IUPAC Name | 1,1,2,2-tetrabromo-1,2-dideuterioethane |
| Molecular Weight (g/mol) | 347.6656 |
| Formula Weight | 343.7016 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction