missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Dichloroethane-2,2,2-d3, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1,1-Dichloroethane 2,2,2-d3 |
| CAS | 56912-77-7 |
| Molecular Formula | C2 D3 H Cl2 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C2H4Cl2/c1-2(3)4/h2H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(Cl)Cl |
| IUPAC Name | 2,2-dichloro-1,1,1-trideuterio-ethane |
| Molecular Weight (g/mol) | 101.98 |
| Formula Weight | 100.9878 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction