missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Dichloroethylene-d2, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1,1-Dichloroethene-d2 |
| CAS | 22280-73-5 |
| Molecular Formula | C2 D2 Cl2 |
| Quantity | 0.1 g |
| Synonym | 1,1-Dichloroethylene-d2 |
| InChI Formula | InChI=1 S/C2H2Cl2/c1-2(3)4/h1H2/i1D2 |
| SMILES | [2 H]C(=C(Cl)Cl)[2 H] |
| IUPAC Name | 1,1-dichloro-2,2-dideuterio-ethylene |
| Molecular Weight (g/mol) | 98.96 |
| Formula Weight | 97.966 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction