missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Dimethyl-d6-urea, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethylurea-D6 |
| CAS | 1219802-32-0 |
| Molecular Formula | C3 D6 H2 N2 O |
| Quantity | 0.1 g |
| Synonym | N,N-Di(methyl-d3)urea, N,N-Dimethylurea-d6, N,N-Dimethylurea-D6, 1,1-Dimethyl-d6-urea, 1,1-Dimethylurea-d6, asym-Dimethylurea-d6, N,N-Di(methyl-d3)carbamide |
| InChI Formula | InChI=1 S/C3H8N2O/c1-5(2)3(4)6/h1-2H3,(H2,4,6)/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])N(C(=O)N)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1-bis(trideuteriomethyl)urea |
| Molecular Weight (g/mol) | 94.1454 |
| Formula Weight | 94.1013 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction