missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Cyclohexane-d10-diamine (cis/trans mixture), CDN
Click to view available options
Quantity:
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | Cyclohexane-1,2-diamine-D10 |
| CAS | 2120097-66-5 |
| Molecular Formula | C6 D10 H4 N2 |
| Quantity | 0.1 g |
| Synonym | 1,2-Cyclohexane-d10-diamine, 1,2-Diaminocyclohexane-d10, Cyclohexane-1,2-diamine-D10 |
| InChI Formula | InChI=1 S/C6H14N2/c7-5-3-1-2-4-6(5)8/h5-6 H,1-4,7-8H2/i1D2,2D2,3D2,4D2,5 D,6 D |
| SMILES | [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(N)C([2 H])(N)C1([2 H])[2 H] |
| IUPAC Name | 1,2,3,3,4,4,5,5,6,6-decadeuteriocyclohexane-1,2-diamine |
| Molecular Weight (g/mol) | 124.2504 |
| Formula Weight | 124.1785 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction