missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Methoxy-d3-phenylazo)-2-naphthol, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-(2-Methoxyphenylazo)-2-naphthol-D3 (methoxy-D3) |
| CAS | 1398109-09-5 |
| Molecular Formula | C17 D3 H11 N2 O2 |
| Quantity | 0.1 g |
| Synonym | 1-(2-Methoxy-d3-phenylazo)-2-naphthol, 1-(2-Methoxyphenylazo)-2-naphthol-D3 (methoxy-D3), 1-[2-[2-(Methoxy-d3)phenyl]diazenyl]-2-naphthalenol, Sudan Red G-d3 (methoxy-d3), Anisole-2-azo-β-naphthol-d3 |
| InChI Formula | InChI=1 S/C17H14N2O2/c1-21-16-9-5-4-8-14(16)18-19-17-13-7-3-2-6-12(13)10-11-15(17)20/h2-11,20 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1ccccc1N=Nc2c(O)ccc3ccccc23 |
| IUPAC Name | 1-[[2-(trideuteriomethoxy)phenyl]diazenyl]naphthalen-2-ol |
| Molecular Weight (g/mol) | 281.3238 |
| Formula Weight | 281.1244 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction