missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Butane-2,2,3,3-d4-diamine 2HCl, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1,4-Diaminobutane-2,2,3,3-D4 Dihydrochloride |
| CAS | 88972-24-1 |
| Molecular Formula | C4H10D4Cl2N2 |
| Quantity | 0.1 g |
| Synonym | 1,4-Butane-2,2,3,3-d4-diamine, dihydrochloride (9 CI), 1,4-Butane-2,2,3,3-d4-diamine 2 HCl, 1,4-Butane-2,2,3,3-d4-diamine dihydrochloride, 1,4-Diaminobutane-2,2,3,3-D4 dihydrochloride, 1,4-Diaminobutane-d4 dihydrochloride, Putrescine-d4 dihydrochloride |
| InChI Formula | InChI=1 S/C4H12N2.ClH/c5-3-1-2-4-6;/h1-6H2;1 H/i1D2,2D2; |
| SMILES | C(C(CN)([2 H])[2 H])(CN)([2 H])[2 H].Cl |
| IUPAC Name | butane-2,2,3,3-d4-1,4-diamine hydrochloride |
| Molecular Weight (g/mol) | 165.09 |
| Formula Weight | 164.08 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction