missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromotetradecane-14,14,14-d3, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromotetradecane-14,14,14-D3 |
| CAS | 347840-09-9 |
| Molecular Formula | C14 D3 H26 Br |
| Quantity | 0.1 g |
| Synonym | Tetradecane-1,1,1-d3, 14-bromo- (9 CI), 14-Bromotetradecane-1,1,1-d3, 1-Bromotetradecane-14,14,14-d3, 1-Bromotetradecane-14,14,14-D3, 1-Tetradecyl-14,14,14-d3 bromide, Myristyl-14,14,14-d3 bromide |
| InChI Formula | InChI=1 S/C14H29Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h2-14H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCCCCCCCCCCCCBr |
| IUPAC Name | 14-bromo-1,1,1-trideuteriotetradecane |
| Molecular Weight (g/mol) | 280.3026 |
| Formula Weight | 279.1641 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction