missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2-Methyl-d3-3-hexanol-1,1,1,2,3,4,4-d7, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-3-hexanol-1,1,1,1',1',1',2,3,4,4-D10 |
| Molecular Formula | C7 D10 H6 O |
| Quantity | 0.1 g |
| Synonym | (±)-2-Methyl-d3-3-hexanol-1,1,1,2,3,4,4-d7, 2-Methyl-3-hexanol-d10, 1-(Isopropyl-d7)-1-butanol-1,2,2-d3, 2-Methyl-3-hexanol-1,1,1,1',1',1',2,3,4,4-D10 |
| InChI Formula | InChI=1 S/C7H16O/c1-4-5-7(8)6(2)3/h6-8 H,4-5H2,1-3H3/i2D3,3D3,5D2,6 D,7 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])(O)C([2 H])([2 H])CC |
| IUPAC Name | 1,1,1,2,3,4,4-heptadeuterio-2-(trideuteriomethyl)hexan-3-ol |
| Molecular Weight (g/mol) | 126.263 |
| Formula Weight | 126.1829 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction