missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenyl-d5-ethyl Isothiocyanate, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Phenethyl Isothiocyanate-D5 (phenyl-D5) |
| CAS | 912627-98-6 |
| Molecular Formula | C9 D5 H4 N S |
| Quantity | 0.1 g |
| Synonym | Benzene-d5, (2-isothiocyanatoethyl)- (9 CI), (2-Isothiocyanatoethyl)benzene-d5, 2-Phenyl-d5-ethyl Isothiocyanate, Phenethyl Isothiocyanate-D5 (phenyl-D5) |
| InChI Formula | InChI=1 S/C9H9NS/c11-8-10-7-6-9-4-2-1-3-5-9/h1-5 H,6-7H2/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CCN=C=S)c([2 H])c1[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-(2-isothiocyanatoethyl)benzene |
| Molecular Weight (g/mol) | 168.2703 |
| Formula Weight | 168.077 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction