missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(N-Morpholino)propanesulfonic Acid-d15, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 3-(N-Morpholino)propanesulfonic Acid-D15 |
| CAS | 1219799-30-0 |
| Molecular Formula | C7 D15 N O4 S |
| Quantity | 0.1 g |
| Synonym | 3-(N-Morpholino)propanesulfonic Acid-d15, 4-Morpholine-d8-propane-1,1,2,2,3,3-d6-sulfonic acid-d, 3-(N-Morpholino-d8)propane-1,1,2,2,3,3-d6-sulfonic acid-d, 3-(N-Morpholino)propanesulfonic Acid-D15, MOPS-d15 |
| InChI Formula | InChI=1 S/C7H15NO4S/c9-13(10,11)7-1-2-8-3-5-12-6-4-8/h1-7H2,(H,9,10,11)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2/hD |
| SMILES | [2 H]OS(=O)(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N1C([2 H])([2 H])C([2 H])([2 H])OC([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | deuterio 1,1,2,2,3,3-hexadeuterio-3-(2,2,3,3,5,5,6,6-octadeuteriomorpholin-4-yl)propane-1-sulfonate |
| Molecular Weight (g/mol) | 224.3557 |
| Formula Weight | 224.1663 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction