missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dimethoxybenzophenone-d8 (rings-d8), CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4,4'-Dimethoxybenzophenone-2,2',3,3',5,5',6,6'-D8 |
| CAS | 350818-55-2 |
| Molecular Formula | C15 D8 H6 O3 |
| Quantity | 0.1 g |
| Synonym | Methanone, bis(4-methoxyphenyl-2,3,5,6-d4)- (9 CI), 4,4'-Dimethoxybenzophenone-d8 (rings-d8), 4,4'-Dimethoxybenzophenone-2,2',3,3',5,5',6,6'-d8, 4,4'-Dimethoxybenzophenone-2,2',3,3',5,5',6,6'-D8, Bis(4-methoxyphenyl-2,3,5,6-d4)methanone |
| InChI Formula | InChI=1 S/C15H14O3/c1-17-13-7-3-11(4-8-13)15(16)12-5-9-14(18-2)10-6-12/h3-10 H,1-2H3/i3D,4 D,5 D,6 D,7 D,8 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)c2c([2 H])c([2 H])c(OC)c([2 H])c2[2 H])c([2 H])c([2 H])c1OC |
| IUPAC Name | bis(2,3,5,6-tetradeuterio-4-methoxyphenyl)methanone |
| Molecular Weight (g/mol) | 250.3192 |
| Formula Weight | 250.1445 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction