missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl-d7-amine HCl, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | Benzylamine-D7 Hydrochloride |
| CAS | 352431-27-7 |
| Molecular Formula | C7 D7 H2 N . Cl H |
| Quantity | 0.1 g |
| Synonym | Benzene-d5-methan-d2-amine, hydrochloride (9 CI), Benzene-d5-methan-d2-amine hydrochloride, Benzyl-d7-amine hydrochloride, Benzylamine-d7 hydrochloride, Benzylamine-D7 hydrochloride |
| InChI Formula | InChI=1 S/C7H9N.ClH/c8-6-7-4-2-1-3-5-7;/h1-5 H,6,8H2;1 H/i1D,2 D,3 D,4 D,5 D,6D2; |
| SMILES | Cl.[2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])N |
| IUPAC Name | dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methanamine;hydrochloride |
| Molecular Weight (g/mol) | 150.6571 |
| Formula Weight | 150.0941 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction