missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biphenyl-4,4'-d2, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Biphenyl-4,4'-D2 |
| CAS | 6120-99-6 |
| Molecular Formula | C12 D2 H8 |
| Quantity | 0.1 g |
| Synonym | 1,1'-Biphenyl-4,4'-d2 (9 CI), Biphenyl-4,4'-d2 (6 CI, 7 CI, 8 CI), 4,4'-d2-Biphenyl, 4,4'-Dideuterobiphenyl, Biphenyl-p,p'-d2, Biphenyl-4,4'-D2 |
| InChI Formula | InChI=1 S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10 H/i1D,2 D |
| SMILES | [2 H]c1ccc(cc1)c2ccc([2 H])cc2 |
| IUPAC Name | 1-deuterio-4-(4-deuteriophenyl)benzene |
| Molecular Weight (g/mol) | 156.2201 |
| Formula Weight | 156.0908 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction