missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1,2,3,6-Tetrahydrophthalimide-3,3,4,5,6,6-d6, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | cis-1,2,3,6-Tetrahydrophthalimide-3,3,4,5,6,6-d6 |
| CAS | 203578-24-9 |
| Molecular Formula | C8 D6 H3 N O2 |
| Quantity | 0.1 g |
| Synonym | cis-1,2,3,6-Tetrahydrophthalimide D6 (3,3,4,5,6,6-D6), 1 H-Isoindole-1,3(2 H)-dione-4,5,6,7-d4, 3 A,4,7,7 A-tetrahydro-4,7-d2-, (3 aR,7 AS)-rel-, 1 H-Isoindole-1,3(2 H)-dione-4,5,6,7-d4, 3 A,4,7,7 A-tetrahydro-4,7-d2-, cis- (ZCI) |
| InChI Formula | InChI=1 S/C8H9NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-2,5-6 H,3-4H2,(H,9,10,11)/t5-,6+/i1D,2 D,3D2,4D2 |
| SMILES | [2 H]C1=C([2 H])C([2 H])([2 H])[C@H]2 [C@H](C(=O)NC2=O)C1([2 H])[2 H] |
| IUPAC Name | rac-(3 aR,7 AS)-4,4,5,6,7,7-hexadeuterio-3 A,7 A-dihydroisoindole-1,3-dione |
| Molecular Weight (g/mol) | 157.2 |
| Formula Weight | 157.101 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction