missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Threonine-2,3-d2, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | D-Threonine-2,3-D2 |
| Molecular Formula | C4 D2 H7 N O3 |
| Quantity | 0.1 g |
| Synonym | D-Threonine-2,3-d2, (2 R,3 S)-2-Amino-2,3-dideuterio-3-hydroxybutanoic acid, D-Threonine-2,3-D2, (2 R,3 S)-2-Amino-3-hydroxybutanoic-2,3-d2 acid |
| InChI Formula | InChI=1 S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6 H,5H2,1H3,(H,7,8)/t2-,3+/m0/s1/i2D,3 D |
| SMILES | [2 H][C@@](C)(O)[C@@]([2 H])(N)C(=O)O |
| IUPAC Name | (2 R,3 S)-2-amino-2,3-dideuterio-3-hydroxybutanoic acid |
| Molecular Weight (g/mol) | 121.1315 |
| Formula Weight | 121.0708 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction