missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Phthalate-3,4,5,6-d4, CDN

Product Code. 30311010
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30311010

Brand: CDN CDND23050.1G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-methyl ester D4
CAS 93951-89-4
Molecular Formula C10 2H4 H6 O4
Quantity 0.1 g
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-dimethyl ester, 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI), Phthalic acid dimethyl ester D4
InChI Formula InChI=1 S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6 H,1-2H3/i3D,4 D,5 D,6 D
SMILES [2 H]c1c([2 H])c([2 H])c(C(=O)OC)c(C(=O)OC)c1[2 H]
IUPAC Name dimethyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molecular Weight (g/mol) 198.21
Formula Weight 198.083 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado