missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Tri(hexadecanoate-13,13,14,14,15,15,16,16,16-d9), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | Tripalmitin-D27 (tri(hexadecanoyl-13,13,14,14,15,15,16,16,16-D9)) |
| CAS | 1219804-91-7 |
| Molecular Formula | C51 D27 H71 O6 |
| Quantity | 0.1 g |
| Synonym | Hexadecanoic-13,13,14,14,15,15,16,16,16-d9 acid 1,2,3-propanetriyl ester, Glyceryl Tri(hexadecanoate-13,13,14,14,15,15,16,16,16-d9), Glycerol trihexadecanoate-d27, Glyceryl tripalmitate-d27, Trihexadecanoin-d27, Tripalmitin-d27, Tripalmitin-D27 (tri(hexadecanoyl-13,13,14,14,15,15,16,16,16-D9)) |
| InChI Formula | InChI=1 S/C51H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h48H,4-47H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])OC(=O)CCCCCCCCCCCC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3-bis(13,13,14,14,15,15,16,16,16-nonadeuteriohexadecanoyloxy)propyl 13,13,14,14,15,15,16,16,16-nonadeuteriohexadecanoate |
| Molecular Weight (g/mol) | 834.4866 |
| Formula Weight | 833.9058 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction