missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Tri(pentadecanoate-d29), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Tripentadecanoin-D87 (tri(pentadecanoyl-D29)) |
| CAS | 352431-43-7 |
| Molecular Formula | C48 D87 H5 O6 |
| Quantity | 0.1 g |
| Synonym | Pentadecanoic-d29 acid, 1,2,3-propanetriyl ester (9 CI), 1,2,3-Tris(pentadecanoyl-d29)glycerol, Tripentadecanoin-d87, Tripentadecanoin-D87 (tri(pentadecanoyl-D29)), Pentadecanoic-d29 acid 1,1',1''-(1,2,3-propanetriyl) ester, Glyceryl Tri(pentadecanoate-d29) |
| InChI Formula | InChI=1 S/C48H92O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-46(49)52-43-45(54-48(51)42-39-36-33-30-27-24-21-18-15-12-9-6-3)44-53-47(50)41-38-35-32-29-26-23-20-17-14-11-8-5-2/h45H,4-44H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2,23D2,24D2,25D2,26D2,27D2,28D2,29D2,30D2,31D2,32D2,33D2,34D2,35D2,36D2,37D2,38D2,39D2,40D2,41D2,42D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OCC(COC(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])OC(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3-bis(2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,15-nonacosadeuteriopentadecanoyloxy)propyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,15-nonacosadeuteriopentadecanoate |
| Molecular Weight (g/mol) | 852.7766 |
| Formula Weight | 852.2355 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction