missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Trihexadecanoate-d98, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Tripalmitin-D98 |
| CAS | 1219803-85-6 |
| Molecular Formula | C51 D98 O6 |
| Quantity | 0.1 g |
| Synonym | Hexadecanoic-d31 acid 1,2,3-propanetriyl-1,1,2,3,3-d5 ester, Glyceryl-d5 Tri(hexadecanoate-d31), Glycerol trihexadecanoate-d98, Glyceryl tripalmitate-d98, Trihexadecanoin-d98, Tripalmitin-d98, Tripalmitin-D98 |
| InChI Formula | InChI=1 S/C51H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h48H,4-47H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2,23D2,24D2,25D2,26D2,27D2,28D2,29D2,30D2,31D2,32D2,33D2,34D2,35D2,36D2,37D2,38D2,39D2,40D2,41D2,42D2,43D2,44D2,45D2,46D2,47D2,48 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OC([2 H])([2 H])C([2 H])(OC(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])OC(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | [1,1,2,3,3-pentadeuterio-2,3-bis(2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-hentriacontadeuteriohexadecanoyloxy)propyl] 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-hentriacontadeuteriohexadecanoate |
| Molecular Weight (g/mol) | 905.9241 |
| Formula Weight | 905.3515 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction