missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-Iodo-4-methyl-d3-benzoate-2,5,6-d3, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Methyl 3-Iodo-4-methyl-d3-benzoate-2,5,6-D3 |
| Molecular Formula | C9 D3 H6 I O2 |
| Quantity | 0.1 g |
| Synonym | Methyl 3-Iodo-4-methyl-d3-benzoate-2,5,6-D3, 3-Iodo-4-methylbenzoic-2,5,6-d3 acid methyl ester, 3-Iodo-4-methylbenzoic Acid Methyl Ester-2,5,6-d3 |
| InChI Formula | InChI=1 S/C9H9IO2/c1-6-3-4-7(5-8(6)10)9(11)12-2/h3-5 H,1-2H3/i3D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)OC)c([2 H])c(I)c1C |
| IUPAC Name | methyl 2,3,6-trideuterio-5-iodo-4-methylbenzoate |
| Molecular Weight (g/mol) | 279.0895 |
| Formula Weight | 278.9836 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction