missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Dodecyl-d25-phosphocholine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Dodecylphosphocholine-D25 (dodecyl-D25) |
| CAS | 861924-55-2 |
| Molecular Formula | C17 D25 H13 N O4 P |
| Quantity | 0.1 g |
| Synonym | Ethanaminium, 2-[[(dodecyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25-oxy)hydroxyphosphinyl]oxy]-N,N,N-trimethyl-, inner salt (ACI), Ethanaminium, 2-[[(dodecyl-d25-oxy)hydroxyphosphinyl]oxy]-N,N,N-trimethyl-, inner salt (9 CI), n-Dodecyl-d25-phosphocholine, 2-[[(Dodecyl-d25-oxy)hydroxyphosphinyl]oxy]-N,N,N-trimethylethanaminium inner salt, Dodecylphosphocholine-D25 (dodecyl-D25) |
| InChI Formula | InChI=1 S/C17H38NO4P/c1-5-6-7-8-9-10-11-12-13-14-16-21-23(19,20)22-17-15-18(2,3)4/h5-17H2,1-4H3/i1D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,16D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OP(=O)([O-])OCC[N+](C)(C)C |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-pentacosadeuteriododecyl 2-(trimethylazaniumyl)ethyl phosphate |
| Molecular Weight (g/mol) | 376.616 |
| Formula Weight | 376.411 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction