missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexadecyltrimethylammonium-d42 Chloride, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Hexadecyltrimethylammonium-D42 Chloride |
| CAS | 2095863-88-8 |
| Molecular Formula | C19 D42 N . Cl |
| Quantity | 0.1 g |
| Synonym | N,N,N-Tri(methyl-d3)-1-hexadecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-d33-aminium chloride, Hexadecyltrimethylammonium chloride-d42, Cetrimonium-d42 chloride, Hexadecyltrimethylammonium-D42 chloride |
| InChI Formula | InChI=1 S/C19H42N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1 H/q+1;/p-1/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[N+](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | tris(trideuteriomethyl)-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-tritriacontadeuteriohexadecyl)azanium;chloride |
| Molecular Weight (g/mol) | 362.255 |
| Formula Weight | 361.564 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction