missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosodi-n-butyl-d18-amine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | N-Nitroso-di-n-butylamine d18 |
| CAS | 1219798-82-9 |
| Molecular Formula | C8 2H18 N2 O |
| Quantity | 0.1 g |
| Synonym | Dibutylnitrosamine-d18, NDBA-d18, NSC 6830-d18, Nitrosodibutylamine-d18, N-Butyl-N-nitroso-1-butanamine-d18 |
| InChI Formula | InChI=1 S/C8H18N2O/c1-3-5-7-10(9-11)8-6-4-2/h3-8H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N(N=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N-bis(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)nitrous amide |
| Molecular Weight (g/mol) | 176.35 |
| Formula Weight | 176.2549 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction