missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluidine-d3 (methyl-d3), CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4-Methylaniline-D3 (methyl-D3) |
| CAS | 23346-25-0 |
| Molecular Formula | C7 D3 H6 N |
| Quantity | 0.1 g |
| Synonym | Benzenamine, 4-(methyl-d3)- (ACI), 4-(Methyl-d3)benzenamine (ACI), p-Toluidine-α,α,α-d3 (8 CI), p-Trideuteriomethylaniline, p-Toluidine-d3 (methyl-d3) |
| InChI Formula | InChI=1 S/C7H9N/c1-6-2-4-7(8)5-3-6/h2-5 H,8H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])c1ccc(N)cc1 |
| IUPAC Name | 4-(trideuteriomethyl)aniline |
| Molecular Weight (g/mol) | 110.1715 |
| Formula Weight | 110.0923 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction