missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium n-Hexadecyl-d33 Sulfate, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Sodium Hexadecyl-D33 Sulfate |
| CAS | 2708287-31-2 |
| Molecular Formula | C16 D33 O4 S . Na |
| Quantity | 0.1 g |
| Synonym | Sodium n-hexadecyl-d33 sulfate, Sulfuric acid monohexadecyl-d33 ester sodium salt, Sodium hexadecyl-D33 sulfate, Sodium hexadecyl sulfate D33 (hexadecyl-D33), Sodium cetyl-d33 sulfate |
| InChI Formula | InChI=1 S/C16H34O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20-21(17,18)19;/h2-16H2,1H3,(H,17,18,19);/q;+1/p-1/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OS(=O)(=O)[O-] |
| IUPAC Name | sodium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-tritriacontadeuteriohexadecyl sulfate |
| Molecular Weight (g/mol) | 377.689 |
| Formula Weight | 377.407 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction