missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetracosanoic-9,9,10,10-d4 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Tetracosanoic-9,9,10,10-D4 Acid |
| CAS | 1219794-61-2 |
| Molecular Formula | C24 D4 H44 O2 |
| Quantity | 0.1 g |
| Synonym | Tetracosanoic-9,9,10,10-d4 acid, Tetracosanoic acid-9,9,10,10-d4 |
| InChI Formula | InChI=1 S/C24H48O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26/h2-23H2,1H3,(H,25,26)/i15D2,16D2 |
| SMILES | [2 H]C([2 H])(CCCCCCCCCCCCCC)C([2 H])([2 H])CCCCCCCC(=O)O |
| IUPAC Name | 9,9,10,10-tetradeuteriotetracosanoic acid |
| Molecular Weight (g/mol) | 372.6614 |
| Formula Weight | 372.3905 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction