missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydro-4H-pyran-4-ol-3,3,4,5,5-d5, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | Tetrahydro-4 H-pyran-4-ol-3,3,4,5,5-D5 |
| CAS | 1219795-46-6 |
| Molecular Formula | C5 D5 H5 O2 |
| Quantity | 0.1 g |
| Synonym | 2 H-Pyran-3,5-d2-4-ol, tetrahydro-3,4,5-d3- (ACI), Tetrahydro-3,4,5-d3-2 H-pyran-3,5-d2-4-ol (ACI), Tetrahydro-4 H-pyran-4-ol-3,3,4,5,5-d5, 4-Hydroxytetrahydro-2 H-pyran-3,3,4,5,5-d5, 4-Hydroxytetrahydropyran-3,3,4,5,5-d5, Tetrahydro-2 H-pyranol-4-ol-3,3,4,5,5-d5, Tetrahydro-2 H-pyranol-4-ol-3,3,4,5,5-d5 |
| InChI Formula | InChI=1 S/C5H10O2/c6-5-1-3-7-4-2-5/h5-6 H,1-4H2/i1D2,2D2,5 D |
| SMILES | [2 H]C1([2 H])COCC([2 H])([2 H])C1([2 H])O |
| IUPAC Name | 3,3,4,5,5-pentadeuteriooxan-4-ol |
| Molecular Weight (g/mol) | 107.1625 |
| Formula Weight | 107.0995 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction