missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tridecanoic-10,10,11,11,12,12,13,13,13-d9 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | Tridecanoic-10,10,11,11,12,12,13,13,13-D9 Acid |
| Molecular Formula | C13 D9 H17 O2 |
| Quantity | 0.1 g |
| Synonym | Tridecanoic acid-d9, Tridecylic-10,10,11,11,12,12,13,13,13-d9 acid, Tridecanoic-10,10,11,11,12,12,13,13,13-D9 acid |
| InChI Formula | InChI=1 S/C13H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13(14)15/h2-12H2,1H3,(H,14,15)/i1D3,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CCCCCCCCC(=O)O |
| IUPAC Name | 10,10,11,11,12,12,13,13,13-nonadeuteriotridecanoic acid |
| Molecular Weight (g/mol) | 223.4 |
| Formula Weight | 223.25 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction