missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trigonelline-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | Trigonelline-d3 Hydrochloric Acid Salt (N-methyl-d3) |
| Molecular Formula | C7 D3 H5 N O2 . Cl |
| Quantity | 0.1 g |
| Synonym | 3-Carboxy-1-(methyl-d3)pyridinium chloride, Trigonelline-d3 hydrochloride, Trigonelline-D3 (N-methyl-D3) chloride |
| InChI Formula | InChI=1 S/C7H7NO2.ClH/c1-8-4-2-3-6(5-8)7(9)10;/h2-5 H,1H3;1 H/i1D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[n+]1 cccc(c1)C(=O)O |
| IUPAC Name | 1-(trideuteriomethyl)pyridin-1-ium-3-carboxylic acid;chloride |
| Molecular Weight (g/mol) | 176.615 |
| Formula Weight | 176.043 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction