Learn More
Merck 0.1KG Used as coating to enhance cell attachment and adhesion to both plasticware and glass surfaces
Brand:  Merck A-005-C
0.1KG Used as coating to enhance cell attachment and adhesion to both plasticware and glass surfaces
Product Code. 17199660
  • 245.00 € / 0.10kg