missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromotetradecane-1,1,2,2-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | 1-Bromotetradecane-1,1,2,2-D4 |
| CAS | 1219798-81-8 |
| Molecular Formula | C14 D4 H25 Br |
| Quantity | 0.25 g |
| Synonym | 1-Bromotetradecane-1,1,2,2-d4, 1-Tetradecyl-1,1,2,2-d4 bromide, Myristyl-1,1,2,2-d4 bromide |
| InChI Formula | InChI=1 S/C14H29Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h2-14H2,1H3/i13D2,14D2 |
| SMILES | [2 H]C([2 H])(Br)C([2 H])([2 H])CCCCCCCCCCCC |
| IUPAC Name | 1-bromo-1,1,2,2-tetradeuteriotetradecane |
| Molecular Weight (g/mol) | 281.3087 |
| Formula Weight | 280.1704 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction