missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Difluorotoluene-3,5,6-d3, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 2,4-Difluorotoluene-3,5,6-D3 |
| CAS | 1219798-79-4 |
| Molecular Formula | C7 D3 H3 F2 |
| Quantity | 0.25 g |
| Synonym | 4,6-Difluoro-1-methylbenzene-2,3,5-d3, 2,4-Difluoro-1-methylbenzene-3,5,6-d3, 2,4-Difluorotoluene-3,5,6-d3, 2,4-Difluorotoluene-3,5,6-D3 |
| InChI Formula | InChI=1 S/C7H6F2/c1-5-2-3-6(8)4-7(5)9/h2-4 H,1H3/i2D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(F)c([2 H])c(F)c1C |
| IUPAC Name | 1,2,4-trideuterio-3,5-difluoro-6-methylbenzene |
| Molecular Weight (g/mol) | 131.1378 |
| Formula Weight | 131.0626 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction