missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethylphenol-d10, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2,4-Dimethylphenol d10 |
| CAS | 1219794-86-1 |
| Molecular Formula | C8 2H10 O |
| Quantity | 0.25 g |
| Synonym | 2,4-Dimethylphenol D10, 2,4-Dimethylphenol-d10, 2,4-Bis(trideuteriomethyl-3,5,6-trideuteriophenol-OD, Perdeuterio-2,4-dimethylphenol |
| InChI Formula | InChI=1 S/C8H10O/c1-6-3-4-8(9)7(2)5-6/h3-5,9 H,1-2H3/i1D3,2D3,3 D,4 D,5 D/hD |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c(c([2 H])c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,5-trideuterio-3-deuteriooxy-4,6-bis(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 132.23 |
| Formula Weight | 132.1359 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction