missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroethyl-d4-amine HCl, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-ethanamine 1,1,2,2-d4 hydrochloride |
| CAS | 172333-26-5 |
| Molecular Formula | C2 2H4 H2 Cl N . Cl H |
| Quantity | 0.25 g |
| Synonym | 2-Chloroethan-1,1,2,2-d4-amine hydrochloride (9 CI) |
| InChI Formula | InChI=1 S/C2H6ClN.ClH/c3-1-2-4;/h1-2,4H2;1 H/i1D2,2D2; |
| SMILES | Cl.[2 H]C([2 H])(N)C([2 H])([2 H])Cl |
| IUPAC Name | 2-chloro-1,1,2,2-tetradeuterioethanamine;hydrochloride |
| Molecular Weight (g/mol) | 120.01 |
| Formula Weight | 119.0207 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction