missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxyphenol-3,4,5,6-d4,OD, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Methoxyphenol-O,3,4,5,6-D5 |
| CAS | 20189-11-1 |
| Molecular Formula | C7 D5 H3 O2 |
| Quantity | 0.25 g |
| Synonym | Phen-2,3,4,5-d4-ol-d, 6-methoxy- (8 CI, 9 CI), 2-Methoxyphen-d4-ol-d, OD-3,4,5,6-Tetradeuteroguaiacol, 2-Methoxyphenol-O,3,4,5,6-d5, Guaiacol-d4 (OD,ring-d4), 2-Methoxyphenol-O,3,4,5,6-D5, 2-Methoxyphenol-d5, 2-Methoxyphenol-3,4,5,6-d4,OD |
| InChI Formula | InChI=1 S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8 H,1H3/i2D,3 D,4 D,5 D/hD |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c([2 H])c([2 H])c1OC |
| IUPAC Name | 1,2,3,4-tetradeuterio-5-deuteriooxy-6-methoxybenzene |
| Molecular Weight (g/mol) | 129.168 |
| Formula Weight | 129.0838 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction