missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenoxyethyl-1,1-d2 Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Phenoxyethanol-d2 |
| CAS | 21273-38-1 |
| Molecular Formula | C8 D2 H8 O2 |
| Quantity | 0.25 g |
| Synonym | Ethan-1,1-d2-ol, 2-phenoxy- (8 CI,9 CI), Phenoxyethanol-d2, (2-Hydroxyethoxy-2,2-d2)benzene, β-Hydroxyethyl-d2 phenyl ether, β-Phenoxyethanol-d2, 1-Hydroxy-2-phenoxyethane-1,1-d2, 2-Hydroxyethyl-d2 phenyl ether, 2-Phenoxyethanol-d2, 2-Phenoxyethyl-1,1-d2 alcohol, 2-(Phenoxy)ethanol-1,1-D2 |
| InChI Formula | InChI=1 S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9 H,6-7H2/i6D2 |
| SMILES | [2 H]C([2 H])(O)COc1ccccc1 |
| IUPAC Name | 1,1-dideuterio-2-phenoxyethanol |
| Molecular Weight (g/mol) | 140.176 |
| Formula Weight | 140.081 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction