missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethyl-d6-aniline-5-d1, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 3,4-Dimethylaniline-D7 (5-D,dimethyl-D6) |
| Molecular Formula | C8 D7 H4 N |
| Quantity | 0.25 g |
| Synonym | 4,5-Di(methyl-d3)benzen-3-d-amine, 3,4-Dimethylaniline-d7, 3,4-Dimethylaniline-D7 (5-D,dimethyl-D6), 3,4-Di(methyl-d3)aniline-5-d1 |
| InChI Formula | InChI=1 S/C8H11N/c1-6-3-4-8(9)5-7(6)2/h3-5 H,9H2,1-2H3/i1D3,2D3,3 D |
| SMILES | [2 H]c1cc(N)cc(c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 3-deuterio-4,5-bis(trideuteriomethyl)aniline |
| Molecular Weight (g/mol) | 128.2228 |
| Formula Weight | 128.1331 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction