missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylbutyraldehyde-2,2-d2, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | 3-Methylbutyraldehyde-2,2-d2 |
| CAS | 352431-47-1 |
| Molecular Formula | C5 D2 H8 O |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C5H10O/c1-5(2)3-4-6/h4-5 H,3H2,1-2H3/i3D2 |
| SMILES | [2 H]C([2 H])(C=O)C(C)C |
| IUPAC Name | 2,2-dideuterio-3-methylbutanal |
| Molecular Weight (g/mol) | 88.145 |
| Formula Weight | 88.086 g/mol |
| Percent Purity | 98 atom % D, min 96% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction