missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylpentanedioic-2,2,4,4-d4 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | 3-Methylpentanedioic Acid-2,2,4,4-D4 |
| CAS | 1219798-68-1 |
| Molecular Formula | C6 D4 H6 O4 |
| Quantity | 0.25 g |
| Synonym | 3-Methylpentanedioic-2,2,4,4-d4 Acid, 3-Methylpentanedioic Acid-2,2,4,4-D4, 3-Methylglutaric acid-2,2,4,4-d4, β-Methylglutaric acid-α,γ-d4 |
| InChI Formula | InChI=1 S/C6H10O4/c1-4(2-5(7)8)3-6(9)10/h4H,2-3H2,1H3,(H,7,8)(H,9,10)/i2D2,3D2 |
| SMILES | [2 H]C([2 H])(C(C)C([2 H])([2 H])C(=O)O)C(=O)O |
| IUPAC Name | 2,2,4,4-tetradeuterio-3-methylpentanedioic acid |
| Molecular Weight (g/mol) | 150.1659 |
| Formula Weight | 150.083 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction