missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylbutyric-d11 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 4-Phenylbutyric acid-d11 |
| CAS | 358730-86-6 |
| Molecular Formula | C10 2H11 H O2 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C10H12O2/c11-10(12)8-4-7-9-5-2-1-3-6-9/h1-3,5-6 H,4,7-8H2,(H,11,12)/i1D,2 D,3 D,4D2,5 D,6 D,7D2,8D2 |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4-hexadeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)butanoic acid |
| Molecular Weight (g/mol) | 175.27 |
| Formula Weight | 175.1528 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction