missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-7-Methyl-d3-nonanoic Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 7-Methylnonanoic Acid-D3 (7-methyl-D3) |
| CAS | 2708280-22-0 |
| Molecular Formula | C10 D3 H17 O2 |
| Quantity | 0.25 g |
| Synonym | 7-Methylnonanoic Acid-D3 (7-methyl-D3), (±)-7-Methyl-d3-nonanoic acid |
| InChI Formula | InChI=1 S/C10H20O2/c1-3-9(2)7-5-4-6-8-10(11)12/h9H,3-8H2,1-2H3,(H,11,12)/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(CC)CCCCCC(=O)O |
| IUPAC Name | 7-(trideuteriomethyl)nonanoic acid |
| Molecular Weight (g/mol) | 175.2831 |
| Formula Weight | 175.1652 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction