missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetamide-d5, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Acetamide-d5 |
| CAS | 33675-83-1 |
| Molecular Formula | C2 D5 N O |
| Quantity | 0.25 g |
| Synonym | Acetamide-d5 (7 CI, 8 CI, 9 CI), Acetamide-D5, Acetamide-N,N,2,2,2-d5 |
| InChI Formula | InChI=1 S/C2H5NO/c1-2(3)4/h1H3,(H2,3,4)/i1D3/hD2 |
| SMILES | [2 H]N([2 H])C(=O)C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N,2,2,2-pentadeuterioacetamide |
| Molecular Weight (g/mol) | 64.098 |
| Formula Weight | 64.0685 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction