missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha,alpha'-Dibromo-p-xylene-d8, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | 1,4-Bis(bromomethyl)benzene-D8 |
| CAS | 74903-77-8 |
| Molecular Formula | C8 D8 Br2 |
| Quantity | 0.25 g |
| Synonym | Benzene-1,2,4,5-d4, 3,6-bis(bromomethyl-d2)- (9 CI, ACI), 3,6-Bis(bromomethyl-d2)benzene-1,2,4,5-d4 (ACI), 1,4-Di(bromomethyl-d2)benzene-2,3,5,6-d4, 1,4-Xylylene-d8 dibromide, α,α'-Dibromo-p-xylene-d8, 1,4-Bis(bromomethyl)benzene-D8 |
| InChI Formula | InChI=1 S/C8H8Br2/c9-5-7-1-2-8(6-10)4-3-7/h1-4 H,5-6H2/i1D,2 D,3 D,4 D,5D2,6D2 |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1C([2 H])([2 H])Br)C([2 H])([2 H])Br |
| IUPAC Name | 1,4-bis[bromo(dideuterio)methyl]-2,3,5,6-tetradeuteriobenzene |
| Molecular Weight (g/mol) | 272.0064 |
| Formula Weight | 269.9495 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction