missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl-2,3,4,5,6-d5 Benzoate, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Benzyl Benzoate-D5 (phenyl-D5-methyl) |
| CAS | 347840-01-1 |
| Molecular Formula | C14 D5 H7 O2 |
| Quantity | 0.25 g |
| Synonym | Benzene-d5-methanol, benzoate (9 CI), Benzoic acid phenyl-d5-methyl ester, Benzyl Benzoate-d5, Benzyl Benzoate-D5 (phenyl-D5-methyl), Benzoic acid benzyl-2,3,4,5,6-d5 ester |
| InChI Formula | InChI=1 S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10 H,11H2/i1D,3 D,4 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(COC(=O)c2ccccc2)c([2 H])c1[2 H] |
| IUPAC Name | (2,3,4,5,6-pentadeuteriophenyl)methyl benzoate |
| Molecular Weight (g/mol) | 217.2747 |
| Formula Weight | 217.1151 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction