missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bisphenol A-3,3',5,5'-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Bisphenol A-3,3',5,5'-d4 |
| CAS | 347841-41-2 |
| Molecular Formula | C15 D4 H12 O2 |
| Quantity | 0.25 g |
| Synonym | Phen-2,6-d2-ol, 4,4'-(1-methylethylidene)bis- (9 CI), Bisphenol A-3,3',5,5'-d4, Bisphenol A-3,3',5,5'-D4, 4,4'-(1-Methylethylidene)bisphen-2,6-d2-ol |
| InChI Formula | InChI=1 S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17 H,1-2H3/i7D,8 D,9 D,10 D |
| SMILES | [2 H]c1cc(cc([2 H])c1O)C(C)(C)c2cc([2 H])c(O)c([2 H])c2 |
| IUPAC Name | 2,6-dideuterio-4-[2-(3,5-dideuterio-4-hydroxyphenyl)propan-2-yl]phenol |
| Molecular Weight (g/mol) | 232.311 |
| Formula Weight | 232.1401 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction