missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caffeine-d3 (7-methyl-d3), CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Caffeine-d3 (7-methyl-d3) |
| CAS | 26351-04-2 |
| Molecular Formula | C8 D3 H7 N4 O2 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])n1cnc2N(C)C(=O)N(C)C(=O)c12 |
| IUPAC Name | 1,3-dimethyl-7-(trideuteriomethyl)purine-2,6-dione |
| Molecular Weight (g/mol) | 197.209 |
| Formula Weight | 197.0992 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction