missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane-d11, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Cyclohexane-D11 |
| CAS | 1520-59-8 |
| Molecular Formula | C6 D11 H |
| Quantity | 0.25 g |
| Synonym | Cyclohexane-1,1,2,2,3,3,4,4,5,5,6-d11 (ACI), Cyclohexane-d11 (6 CI, 7 CI, 8 CI, 9 CI), Undecadeuteriocyclohexane, Undecadeuterocyclohexane, Cyclohexane-D11 |
| InChI Formula | InChI=1 S/C6H12/c1-2-4-6-5-3-1/h1-6H2/i1D,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C1C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6-undecadeuteriocyclohexane |
| Molecular Weight (g/mol) | 95.2273 |
| Formula Weight | 95.1629 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction