missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Lysine-4,4,5,5-d4 2HCl, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | DL-Lysine-4,4,5,5-D4 Dihydrochloride |
| CAS | 284664-88-6 |
| Molecular Formula | C6 D4 H10 N2 O2 . 2 Cl H |
| Quantity | 0.25 g |
| Synonym | Lysine-4,4,5,5-d4, dihydrochloride (9 CI), DL-Lysine-4,4,5,5-d4 hydrochloride (1:2), DL-Lysine-4,4,5,5-D4 dihydrochloride, DL-Lysine-d4 dihydrochloride, 2,6-Diamino-4,4,5,5-tetradeuteriohexanoic acid dihydrochloride, DL-Lysine-4,4,5,5-d4 2 HCl, DL-Lysine-4,4,5,5-d4 2 HCl |
| InChI Formula | InChI=1 S/C6H14N2O2.2 ClH/c7-4-2-1-3-5(8)6(9)10;;/h5H,1-4,7-8H2,(H,9,10);2*1 H/i1D2,2D2;; |
| SMILES | Cl.Cl.[2 H]C([2 H])(CN)C([2 H])([2 H])CC(N)C(=O)O |
| IUPAC Name | 2,6-diamino-4,4,5,5-tetradeuteriohexanoic acid;dihydrochloride |
| Molecular Weight (g/mol) | 223.1341 |
| Formula Weight | 222.084 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction