missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumaric-2,3-d2 Acid, CDN
Click to view available options
Quantity:
0.25 g
Unit Size:
0.25g
Specifications
Specifications
| Chemical Name or Material | Fumaric Acid-d2 |
| CAS | 24461-32-3 |
| Molecular Formula | C4 D2 H2 O4 |
| Quantity | 0.25 g |
| Synonym | (2 E)-2-Butenedioic Acid-d2, (2 E)-But-2-enedioic Acid-d2, (E)-2-Butenedioic Acid-d2, 2-(E)-Butenedioic Acid-d2, (E)-Butenedioic Acid-d2, Allomaleic Acid-d2, Bakeshure 451-d2, Bakeshure 470-d2, Boletic Acid-d2, FC 33-d2, Lichenic Acid-d2, NSC 2752-d2, Petrom-d2, trans-1,2-Ethylenedicarboxylic Acid-d2, trans-2-Butenedioic Acid-d2, trans-Butenedioic Acid-d2; |
| InChI Formula | InChI=1 S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2 H,(H,5,6)(H,7,8)/b2-1+/i1D,2 D |
| SMILES | [2 H]\C(=C(\[2 H])/C(=O)O)\C(=O)O |
| IUPAC Name | (E)-2,3-dideuteriobut-2-enedioic acid |
| Molecular Weight (g/mol) | 118.08 |
| Formula Weight | 118.0235 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction